Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nurr1/NGFI-B beta/NR4A2 Antibody (HL1852) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB295035 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295035 Supplier Novus Biologicals Supplier No. NBP325582
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Nurr1/NGFI-B beta/NR4A2 Monoclonal antibody specifically detects Nurr1/NGFI-B beta/NR4A2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Nurr1/NGFI-B beta/NR4A2
Applications Immunocytochemistry
Classification Monoclonal
Clone HL1852
Conjugate Unconjugated
Dilution Immunocytochemistry/ Immunofluorescence 1:100-1:1000
Formulation PBS
Gene Alias human homolog of, nuclear receptor subfamily 4, group A, member 2, orphan nuclear receptor NR4A2, Orphan nuclear receptor NURR1, T-cell nuclear receptor NOT, transcriptionally inducible nuclear receptor related 1, Transcriptionally-inducible nuclear receptor
Host Species Rabbit
Immunogen Recombinant fragment of human Nurr1/NGFI-B beta/NR4A2.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4929
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.