Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUT Antibody, Novus Biologicals™
SDP

Catalog No. NBP231370 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP231370 0.1 mg
NBP231370SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP231370 Supplier Novus Biologicals Supplier No. NBP231370
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NUT Polyclonal specifically detects NUT in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen NUT
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 0.01 - 0.1 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q86Y26
Gene Alias C15orf55, chromosome 15 open reading frame 55, DKFZp434O192, FAM22H, MGC138683, MGC138684, Nuclear protein in testis, NUT, protein NUT
Gene Symbols NUTM1
Host Species Rabbit
Immunogen Partial recombinant human NUT protein (between residues 400-700) [UniProt Q86Y26[
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256646
Test Specificity Human. Immunogen sequence shows 74% similarity with Mouse NUT protein.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.