Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUT Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
Antigen | NUT |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 0.01 - 0.1 ug/ml |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP231370SS
![]() |
Novus Biologicals
NBP231370SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP231370
![]() |
Novus Biologicals
NBP231370 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
NUT Polyclonal specifically detects NUT in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
NUT | |
Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 0.01 - 0.1 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS with 0.05% Sodium Azide | |
C15orf55, chromosome 15 open reading frame 55, DKFZp434O192, FAM22H, MGC138683, MGC138684, Nuclear protein in testis, NUT, protein NUT | |
NUTM1 | |
IgG | |
Protein A purified | |
Human. Immunogen sequence shows 74% similarity with Mouse NUT protein. |
1.0 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
Q86Y26 | |
256646 | |
Partial recombinant human NUT protein (between residues 400-700) [UniProt Q86Y26[ | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title