Learn More
Description
Biological Activity: MDM2 antagonist; inhibits the MDM2-p53 interaction (IC50 = 0.09 µM) and activates p53. Antiproliferative agent; induces apoptosis in cancer cells.
Storage: Store at 4°C.
Note: Inhibitors are guaranteed for 1 year from date of receipt.
Specifications
Specifications
| Color | White |
| Product Type | Inhibitor |
| Target Proteins | Nutlin-3 |
| CAS | 548472-68-0 |
| Molecular Formula | C30H30Cl2N4O4 |
| Formula Weight | 581.49g/mol |
| Solubility | Soluble to 100mM in DMSO, 100mM in ethanol |
| Content And Storage | Store at 4°C |
| For Use With (Application) | Inhibits apoptosis |
| Quantity | 50 mg |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
