Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Nutlin 3a
GREENER_CHOICE

Catalog No. p-7116097
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

MDM2 antagonist; active enantiomer of Nutlin-3 (Cat. No. 3984)

Nutlin 3a is an MDM2 antagonist (IC50 = 90 nM); inhibits the MDM2-p53 interaction. Active enantiomer of Nutlin-3 (Cat. No. 3984). Induces expression for P53 regulated genes. Suppresses tumor growth in vivo >98% in prostate cancer and osteosarcoma cancer models. Antiproliferative.

Specifications

Chemical Name or Material 4-[[(4S,5R)-4,5-Bis(4-chlorophenyl)-4,5-dihydro-2-[4-methoxy-2-(1-methylethoxy)phenyl]-1H-imidazol-1-yl]carbonyl]-2-piperazinone
CAS 675576-98-4
Assay Percent Range >97%
Quantity 25 mg
Molecular Formula C30H30Cl2N4O4
Formula Weight Observed MW: 581.49
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.