Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NVS PAK1 1
GREENER_CHOICE

Catalog No. p-7116102
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-322-5 25 mg
61-325 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-322-5 Supplier Tocris Bioscience™ Supplier No. 6132/25
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective PAK1 inhibitor

NVS PAK1 1 is a potent and selective PAK1 inhibitor (IC50 = 5 nM) that displays selective for PAK1 over PAK2 and a panel of 442 kinases. NVS PAK1 1 inhibits autophosphorylation of PAK1 in KRAS-mutated pancreatic cancer cell lines.

A negative control for NVS PAK1 1 is also available; NVS PAK1 C .

Specifications

Chemical Name or Material (3S)-3-(2-Chloro-5-(2,2-difluoroethyl)-8-fluoro-5H-dibenzo[b,e][1,4]diazepin-11-ylamino)-N-isopropylpyrrolidine-1-carboxamide
CAS 1783816-74-9
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C23H25ClF3N5O
Formula Weight Observed MW: 479.93
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.