Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NXF1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB180888 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB180888 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB180888 Supplier Novus Biologicals Supplier No. NBP306455AF532
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NXF1 Polyclonal antibody specifically detects NXF1 in Human samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen NXF1
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias DKFZp667O0311, Mex67, mRNA export factor TAP, nuclear RNA export factor 1, TAPMEX67, tip associating protein, Tip-associated protein, Tip-associating protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NXF1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10482
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.