Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NXNL2 Antibody (OTI2B11), HRP, Novus Biologicals™
SDP

Catalog No. NBP273134H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273134H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273134H Supplier Novus Biologicals Supplier No. NBP273134H

Mouse Monoclonal Antibody

NXNL2 Monoclonal specifically detects NXNL2 in Human, Mouse, Monkey, Primate samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen NXNL2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2B11
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias C9orf121, chromosome 9 open reading frame 121, nucleoredoxin-like 2, nucleoredoxin-like protein 2, RDCVF2
Gene Symbols NXNL2
Host Species Mouse
Immunogen Full length recombinant protein of human NXNL2 (NP_660326) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 158046.0
Target Species Human, Mouse, Monkey, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.