Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NXT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | NXT2 |
---|---|
Concentration | 0.16 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7741
![]() ![]() |
Proteintech Group Inc
121361AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Regulator of protein export for NES-containing proteins. Also plays a role in mRNA nuclear export.Specifications
NXT2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
B2GV77, Q3UNA4, Q9NPJ8 | |
237082, 315352, 55916 | |
NXT2 Fusion Protein Ag2782 | |
Primary | |
-20°C | |
NXT2 |
0.16 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DC9, NTF2 related export protein 2, NXT2, P15 2, Protein p15 2 | |
Nxt2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title