Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Anisidine, 98+%

Catalog No. AC104810010
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle

CAS: 90-04-0 | C7H9NO | 123.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-04-0
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.15
MDL Number MFCD00007688
InChI Key VMPITZXILSNTON-UHFFFAOYSA-N
Synonym o-anisidine, 2-anisidine, ortho-anisidine, 2-aminoanisole, o-methoxyaniline, o-aminoanisole, benzenamine, 2-methoxy, 2-methoxybenzenamine, o-methoxyphenylamine, o-anisylamine
PubChem CID 7000
ChEBI CHEBI:82288
IUPAC Name 2-methoxyaniline
SMILES COC1=CC=CC=C1N

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 3.0°C to 6.0°C
Density 1.0920g/mL
Boiling Point 225.0°C
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5730 to 1.5750
Linear Formula CH3OC6H4NH2
Quantity 1 L
Beilstein 13, 358
Specific Gravity 1.092
Merck Index 14, 667
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: soluble in dilute mineral acid, miscible with alcohol and ether, soluble in acetone and benzene
Formula Weight 123.15
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material o-Anisidine, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing genetic defects.
May cause cancer.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-963-1
RTECSNumber BZ5410000
TSCA TSCA
Recommended Storage This substance darkens on exposure to air

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.