Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-GlcNAc Antibody (HGAC85), Novus Biologicals™
SDP

Catalog No. NB300614SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300614SS 0.025 mL
NB300614 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300614SS Supplier Novus Biologicals Supplier No. NB300614SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

O-GlcNAc Monoclonal specifically detects O-GlcNAc in Human, All Species, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen O-GlcNAc
Applications Western Blot, ChIP Assay, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone HGAC85
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Chromatin Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 20368426), ELISA 1:100-1:2000, Immunohistochemistry 1:200. Use reported in scientific literature (Zhang, et al), Immunocytochemistry/Immunofluorescence 1:100 - 1:200. Use reported in scientific literature (Hamiel CR et al), Immunoprecipitation 1:10-1:500. Use reported in scientific literature (Hamiel CR et al), Immunohistochemistry-Paraffin 1:200, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Formulation Ascites with 0.1% Sodium Azide
Gene Alias GlcNAc, O-linked N-acetylglucosamine
Host Species Mouse
Immunogen Heat-killed, pepsin-treated group A streptococci (Streptococcus pyogenes)
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cell Biology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Test Specificity Recognizes beta-1,3 linked O-linked N-acetylglucosamine (O-GlcNAc) residues of streptococcal group A carbohydrate as well as O-GlcNAc glycosylated proteins
Target Species Human, All species, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG3 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.