Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
O-GlcNAc Antibody (HGAC85), Novus Biologicals™

Mouse Monoclonal Antibody
$192.47 - $517.59
Specifications
| Antigen | O-GlcNAc |
|---|---|
| Clone | HGAC85 |
| Dilution | Western Blot 1:500-1:1000, Chromatin Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 20368426), ELISA 1:100-1:2000, Immunohistochemistry 1:200. Use reported in scientific literature (Zhang, et al), Immunocytochemistry/Immunofluorescence 1:100 - 1:200. Use reported in scientific literature (Hamiel CR et al), Immunoprecipitation 1:10-1:500. Use reported in scientific literature (Hamiel CR et al), Immunohistochemistry-Paraffin 1:200, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
| Applications | Western Blot, ChIP Assay, ELISA, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB300614SS
![]() |
Novus Biologicals
NB300614SS |
0.025 mL |
Each for $192.47
|
|
|||||
NB300614
![]() |
Novus Biologicals
NB300614 |
0.1 mL |
Each for $517.59
|
|
|||||
Description
O-GlcNAc Monoclonal specifically detects O-GlcNAc in Human, All Species, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| O-GlcNAc | |
| Western Blot 1:500-1:1000, Chromatin Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 20368426), ELISA 1:100-1:2000, Immunohistochemistry 1:200. Use reported in scientific literature (Zhang, et al), Immunocytochemistry/Immunofluorescence 1:100 - 1:200. Use reported in scientific literature (Hamiel CR et al), Immunoprecipitation 1:10-1:500. Use reported in scientific literature (Hamiel CR et al), Immunohistochemistry-Paraffin 1:200, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, All species, Primate | |
| GlcNAc, O-linked N-acetylglucosamine | |
| IgG3 κ | |
| Unpurified | |
| Recognizes beta-1,3 linked O-linked N-acetylglucosamine (O-GlcNAc) residues of streptococcal group A carbohydrate as well as O-GlcNAc glycosylated proteins |
| HGAC85 | |
| Western Blot, ChIP Assay, ELISA, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Neurodegeneration, Neuroscience, Signal Transduction | |
| Ascites with 0.1% Sodium Azide | |
| Heat-killed, pepsin-treated group A streptococci (Streptococcus pyogenes) | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title