Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-GlcNAc Antibody (RL2), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB300524W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300524W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300524W Supplier Novus Biologicals Supplier No. NB300524AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

O-GlcNAc Monoclonal specifically detects O-GlcNAc in Human, Mouse, Rat, Porcine, Bovine, Drosophila, Fish, Hamster, Primate, Virus, Xenopus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen O-GlcNAc
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RL2
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Gene Alias GlcNAc, O-linked N-acetylglucosamine
Host Species Mouse
Immunogen Pore complex-lamina fraction purified from rat liver nuclear envelopes.
Molecular Weight of Antigen 117 kDa
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects nuclear pore complex (NPC), cytoplasmic and intranuclear O-linked glycoproteins from human, mouse, and rat tissues.
Target Species Human, Mouse, Rat, Pig, Bovine, Drosophila, Fish, Hamster, Primate, Virus, Xenopus species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.