Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
O-GlcNAc Monoclonal Antibody (CTD110.6), Active Motif™

Mouse Monoclonal Antibody
Supplier: Active Motif 61453

Description
O-linked N-acetylglucosamine (O-GlcNAc) is a posttranslational modification characterized by the attachment of N-acetylglucosamine to specific serine or threonine residues. Unlike other protein glycosylations, O-GlcNAc modifications occur within the nucleus and cytoplasm. They are found on many cellular proteins, including nuclear pore, oncogene, cytoskeletal, heat shock, viral and transcription regulatory proteins. O-GlcNAc glycosylations are thought to obscure phosphorylation sites, counteracting phosphorylation-dependent signaling pathways and protein interactions.Specifications
O-GlcNAc | |
Monoclonal | |
1 mg/mL | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
Mouse | |
IgM | |
RUO | |
Human, Multi-species, Mouse, Rat | |
Antibody | |
IgM |
Immunoprecipitation, Western Blot | |
CTD110.6 | |
Unconjugated | |
GlcNAc | |
This antibody was raised against a peptide containing O-GlcNAc - serine. This antibody recognizes single O-GlcNAc residues in b-O-glycosidic linkage to serine and threonine. | |
100 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction