Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-GlcNAc Monoclonal Antibody (CTD110.6), Active Motif™
SDP

Mouse Monoclonal Antibody

$135.30 - $405.90

Specifications

Antigen O-GlcNAc
Clone CTD110.6
Concentration 1 mg/mL
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3411
SDP
Encompass
View Documents
Active Motif
61454
10 μg
Each for $135.30
Only null left
Add to Cart
 
50-199-3410
SDP
Encompass
View Documents
Active Motif
61453
100 μg
Each for $405.90
Only null left
Add to Cart
 
Description

Description

O-linked N-acetylglucosamine (O-GlcNAc) is a posttranslational modification characterized by the attachment of N-acetylglucosamine to specific serine or threonine residues. Unlike other protein glycosylations, O-GlcNAc modifications occur within the nucleus and cytoplasm. They are found on many cellular proteins, including nuclear pore, oncogene, cytoskeletal, heat shock, viral and transcription regulatory proteins. O-GlcNAc glycosylations are thought to obscure phosphorylation sites, counteracting phosphorylation-dependent signaling pathways and protein interactions.
Specifications

Specifications

O-GlcNAc
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
This antibody was raised against a peptide containing O-GlcNAc - serine. This antibody recognizes single O-GlcNAc residues in b-O-glycosidic linkage to serine and threonine.
Primary
-20°C, Avoid Freeze/Thaw Cycles
CTD110.6
Immunoprecipitation, Western Blot
Unconjugated
Mouse
Human, Multi-species, Mouse, Rat
GlcNAc
IgM
IgM
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.