Learn More
O-GlcNAc Monoclonal Antibody (RL2), PE, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
$449.00
Specifications
Antigen | O-GlcNAc |
---|---|
Clone | RL2 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
12-979-342
![]() ![]() |
Invitrogen™
12979342 |
100 Tests |
Each of 1 for $449.00
|
|
|||||
Description
Description: This RL2 monoclonal antibody recognizes proteins with O-N-acetylglucosamine (O-GlcNAc) glycosylation. It was originally developed by immunizing mice with the rat liver nuclear envelopes containing nuclear pore complexes. The RL2 clone has been successfully used in Western blot, immunofluorescence, immunoprecipitation, and flow cytometry in a wide variety of mammalian cells. Applications Reported: This RL2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This RL2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells using the Foxp3/Transcription Factor Staining Buffer Set (Product # 00-5523) and protocol. Please refer to Best Protocols: Protocol B: One step protocol for (nuclear) intracellular proteins located under the Resources Tab online. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
O-linked N-acetylglucosamine (O-GlcNAc) is a posttranslational modification characterized by the attachment of N-acetylglucosamine to specific serine or threonine residues. Unlike other protein glycosylations, O-GlcNAc modifications occur within the nucleus and cytoplasm. They are found on many cellular proteins, including nuclear pore, oncogene, cytoskeletal, heat shock, viral and transcription regulatory proteins. O-GlcNAc glycosylations are thought to obscure phosphorylation sites, counteracting phosphorylation-dependent signaling pathways and protein interactions.Specifications
O-GlcNAc | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
8473 | |
Rat Liver nuclear envelopes. | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
OGT |
RL2 | |
Flow Cytometry | |
PE | |
Mouse | |
O15294 | |
OGT | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.