Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-fluorobenzoate, 99%

Catalog No. AC146801000
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 451-46-7 | C9H9FO2 | 168.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 451-46-7
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.17
MDL Number MFCD00000351
InChI Key UMPRJGKLMUDRHL-UHFFFAOYSA-N
Synonym 4-fluorobenzoic acid ethyl ester, ethyl p-fluorobenzoate, benzoic acid, 4-fluoro-, ethyl ester, p-fluorobenzoic acid, ethyl ester, ethyl4-fluorobenzoate, 4-fluoro-benzoic acid ethyl ester, benzoic acid, p-fluoro-, ethyl ester, ethyep-fluorobenzoate, pubchem3471, ethyl-4-fluorobenzoate
PubChem CID 67976
IUPAC Name ethyl 4-fluorobenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)F

Specifications

Melting Point 25°C to 27°C
Color Colorless to Yellow
Density 1.1400g/mL
Boiling Point 210°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4850 to 1.487
Linear Formula FC6H4CO2C2H5
Quantity 100 g
Beilstein 09, 334
Specific Gravity 1.14
Merck Index 10, 4076
Solubility Information Solubility in water: insoluble
Formula Weight 168.17
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material Ethyl 4-fluorobenzoate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 207-194-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.