Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OAcGD3 Antibody (7H2), Novus Biologicals™
SDP

Catalog No. NB1206218 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1206218 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1206218 Supplier Novus Biologicals Supplier No. NB1206218
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OAcGD3 Monoclonal specifically detects OAcGD3 in Human, Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen OAcGD3
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 7H2
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:10-1:100, Flow Cytometry, ELISA 1:10-1:100
Gene Alias 9-O-acetylated GD3, OAc-GD3
Host Species Mouse
Immunogen O-AcGD3 ganglioside
Purification Method Unpurified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This reacts specifically with O-AcGD3 ganglioside: OAcNeuAc
Reconstitution Reconstitute with 1.0 ml distilled water.
Target Species Non-species specific
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG3 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.