Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OAS2 Antibody (OTI6E6), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP273137V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273137V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273137V Supplier Novus Biologicals Supplier No. NBP273137V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OAS2 Monoclonal specifically detects OAS2 in Human samples. It is validated for Western Blot.

Specifications

Antigen OAS2
Applications Western Blot
Classification Monoclonal
Clone OTI6E6
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias (2-5′)oligo(A) synthase 2, (2′-5′)oligo(A) synthetase 2, (2-5′)oligo(A) synthetase 2, 2′5′ oligoadenylate synthetase 2, 2-5A synthase 2, 2-5A synthetase 2, 2′-5′-oligoadenylate synthase 2, 2′-5′-oligoadenylate synthetase 2 (69-71 kD), 2′-5′-oligoadenylate synthetase 2, 69/71kDa, EC 2.7.7, EC 2.7.7.-, MGC78578, p69 OAS/ p71 OAS, p69OAS/ p71OAS
Gene Symbols OAS2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 60-335 of human OAS2 (NP_002526) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4939.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.