Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OASL Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB191834 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB191834 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB191834 Supplier Novus Biologicals Supplier No. NBP281833AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OASL Polyclonal antibody specifically detects OASL in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen OASL
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias 2'-5'-oligoadenylate synthetase-like, p59 OASL, p59OASL59 kDa 2'-5'-oligoadenylate synthase-like protein, Thyroid receptor-interacting protein 14,59 kDa 2'-5'-oligoadenylate synthetase-like protein, TRIP-14, TRIP14TR-interacting protein 14
Host Species Rabbit
Immunogen OASL antibody was raised against a 17 amino acid peptide near the amino terminus of human OASL. The immunogen is located within the first 50 amino acids of OASL. Amino Acid Squence: VAQWLQPHREWKEEVLD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 8638
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.