Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OAT Antibody (23A2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP149312 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149312 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149312 Supplier Novus Biologicals Supplier No. NBP149312
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OAT Monoclonal specifically detects OAT in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen OAT
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 23A2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:250-1:500, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:250
Gene Accession No. NP_000265
Gene Alias GACR, HOGA, OAT ornithine aminotransferase, OATASE, OKT
Gene Symbols OAT
Host Species Mouse
Immunogen Recombinant human OAT (33-439aa) purified from E. coli.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism, Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 4942
Test Specificity Human OAT (33-439aa).
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.