Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OAT Antibody (23A2), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NBP49312549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP49312549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP49312549 Supplier Novus Biologicals Supplier No. NBP149312JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OAT Monoclonal specifically detects OAT in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen OAT
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 23A2
Conjugate Janelia Fluor 549
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias GACR, HOGA, OAT ornithine aminotransferase, OATASE, OKT
Gene Symbols OAT
Host Species Mouse
Immunogen Recombinant human OAT (33-439aa) purified from E. coli.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline metabolism, Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 4942
Test Specificity Human OAT (33-439aa).
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.