Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OATP1B1/OATP2 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB164637 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164637 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB164637 Supplier Novus Biologicals Supplier No. NBP281750AF405
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OATP1B1/OATP2 Polyclonal antibody specifically detects OATP1B1/OATP2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen OATP1B1/OATP2
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias Liver-specific organic anion transporter 1, LST1SLC21A6, LST-1Solute carrier family 21 member 6, OATP1B1Sodium-independent organic anion-transporting polypeptide 2, OATP2MGC133282, OATP-COATP-2, OATPCsolute carrier organic anion transporter family member 1B1, solute carrier family 21 (organic anion transporter), member 6, solute carrier organic anion transporter family, member 1B1
Host Species Rabbit
Immunogen OATP1B1 antibody was raised against a 14 amino acid peptide near the carboxy terminus of human OATP1B1. The immunogen is located within the last 50 amino acids of OATP1B1. Amino Acid Squence: MKKKYQEKDINASE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 10599
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.