Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OATP2/8 Antibody (MDQ), Novus Biologicals™
SDP

Catalog No. NB10074482 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074482 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074482 Supplier Novus Biologicals Supplier No. NB10074482
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 13 publications

OATP2/8 Monoclonal specifically detects OATP2/8 in Human samples. It is validated for IF/IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen OATP2/8
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MDQ
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:20, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:20, Immunohistochemistry-Frozen 1:20
Formulation Tissue culture supernatant with 0.09% Sodium Azide
Gene Accession No. Q9Y6L6
Gene Alias HBLRR, LST1, LST-1, LST-2, LST3, LST-3TM13, OATP1B1, OATP1B3, OATP2, OATP8, OATP-8, OATPC, OATP-C, SLC21A6, SLC21A8, SLCO1B1 solute carrier organic anion transporter family, member 1B1
Gene Symbols SLCO1B1
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues M(1) D Q N Q H L N K T A E A Q P S E N K K T R Y(23) C of human OATP2.
Purification Method Unpurified
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10599
Test Specificity NB100-74482 detects OATP2 and OATP8 from human tissue and cells.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.