Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OBFC1 Antibody (OTI2E4), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP273138FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273138FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273138FR Supplier Novus Biologicals Supplier No. NBP273138FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OBFC1 Monoclonal specifically detects OBFC1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen OBFC1
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2E4
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias AAF44, AAF-44, alpha accessory factor 44, bA541N10.2, CST complex subunit STN1, oligonucleotide/oligosaccharide-binding fold containing 1, Oligonucleotide/oligosaccharide-binding fold-containing protein 1, replication protein A 32 kDa subunit, RP11-541N10.2, RPA-32, STN1FLJ22559, Suppressor of cdc thirteen homolog
Gene Symbols STN1
Host Species Mouse
Immunogen Full length recombinant protein of human OBFC1 (NP_079204) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 79991.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.