Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OBFC1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB317730 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB317730 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB317730 Supplier Novus Biologicals Supplier No. NBP299011PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OBFC1 Polyclonal antibody specifically detects OBFC1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen OBFC1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias AAF44, AAF-44, alpha accessory factor 44, bA541N10.2, CST complex subunit STN1, oligonucleotide/oligosaccharide-binding fold containing 1, Oligonucleotide/oligosaccharide-binding fold-containing protein 1, replication protein A 32 kDa subunit, RP11-541N10.2, RPA-32, STN1FLJ22559, Suppressor of cdc thirteen homolog
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human OBFC1.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 79991
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.