Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OBFC2B Antibody (OTI1E11), HRP, Novus Biologicals™
SDP

Catalog No. NBP273140H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273140H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273140H Supplier Novus Biologicals Supplier No. NBP273140H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OBFC2B Monoclonal specifically detects OBFC2B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen OBFC2B
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1E11
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias LP3587, MGC2731, oligonucleotide/oligosaccharide-binding fold containing 2B, Oligonucleotide/oligosaccharide-binding fold-containing protein 2B, Sensor of single-strand DNA complex subunit B1, Sensor of ssDNA subunit B1, Single-stranded DNA-binding protein 1, SOSS complex subunit B1, SOSS-B1, SSB1hSSB1
Gene Symbols NABP2
Host Species Mouse
Immunogen Full length recombinant protein of human OBFC2B (NP_076973) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 79035.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.