Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OBP2B Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB351031 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB351031 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB351031 Supplier Novus Biologicals Supplier No. NBP306620MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OBP2B Polyclonal antibody specifically detects OBP2B in Human samples. It is validated for ELISA

Specifications

Antigen OBP2B
Applications ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias hOBPIIb, MGC119022, OBPIIb, odorant binding protein 2B, odorant-binding protein 2b, odorant-binding protein IIb, UNQ653/PRO1283
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human OBP2B (Accession#: NP_055396.1; Met1-His170)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29989
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.