Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OBSCN Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB043270 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB043270 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB043270 Supplier Novus Biologicals Supplier No. NBP297423H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OBSCN Polyclonal specifically detects OBSCN in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen OBSCN
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ARHGEF30, DKFZp666E245, EC 2.7.11.1, FLJ14124, KIAA1556MGC120412, KIAA1639MGC138590, MGC120409, MGC120410, MGC120411, obscurin, obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF, obscurin, myosin light chain kinase, Obscurin-MLCK, Obscurin-myosin light chain kinase, obscurin-RhoGEF, UNC89
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human OBSCN fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 84033
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.