Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Occludin Mouse anti-Human, Clone: OCLN/2181, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30713720UG
Description
Occludin Monoclonal specifically detects Occludin in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
Occludin | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
BLCPMG, occludin | |
Recombinant fragment of human Occludin protein (around aa 282-415) (exact sequence is proprietary) | |
20 μg | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry (Paraffin) | |
OCLN/2181 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
100506658 | |
Store at 4°C. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction