Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Occludin Mouse anti-Human, Clone: OCLN/2181, Novus Biologicals™

Mouse Monoclonal Antibody
$280.00
Specifications
Antigen | Occludin |
---|---|
Clone | OCLN/2181 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Description
Occludin Monoclonal specifically detects Occludin in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
Occludin | |
0.2 mg/ml | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
BLCPMG, occludin | |
Recombinant fragment of human Occludin protein (around aa 282-415) (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
OCLN/2181 | |
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
100506658 | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title