Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Occludin Recombinant Rabbit Monoclonal Antibody (BLR126H)

Rabbit Recombinant Monoclonal Antibody
$190.00 - $459.00
Specifications
| Antigen | Occludin |
|---|---|
| Clone | BLR126H |
| Concentration | 100 μg/mL |
| Content And Storage | 4°C |
| Applications | Immunoprecipitation, Western Blot |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544441
![]() |
Invitrogen™
MA544441 |
100 μL |
Each for $459.00
|
|
|||||
|
PIMA544442
|
Invitrogen™
MA544442 |
20 μL |
Each for $190.00
|
|
|||||
Description
Occludin Recombinant Monoclonal Antibody for Western Blot, IP
Occludin is a 65 kDa protein that can exist in a variety of phosphorylated forms, ranging up to approximately 82 kDa. Occludin is thought to be involved in regulating both the localization and the function of occludin. Polyunsaturated fatty acids are known to up-regulate occludin expression, increasing the transendothelial cell resistance and reducing the cellular permeability to large molecules. The level of occludin varies greatly depending on tissue; in brain tissue, occludin is highly and continuously expressed at cell-cell contact sites, whereas non-neural tissues show lower expression and discontinuous distribution. Overall structural features of the occludin protein are highly conserved in all the species examined. Under-expression of tight junction proteins, including occludin, is a key molecular abnormality responsible for the increased permeability of tumor endothelial tight junctions, which contributes to brain tumor edemas.Specifications
| Occludin | |
| 100 μg/mL | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6 | |
| Q16625 | |
| 100506658 | |
| Peptide representing a region between residue 472 and the C-terminus (residue 522) of Human occludin. | |
| Antibody |
| BLR126H | |
| 4°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| OCLN | |
| AI503564; BLCPMG; Occludin; occludin 1B; Ocl; Ocln; phosphatase 1, regulatory subunit 115; PPP1R115; tight junction protein occludin; tight junction structural protein | |
| OCLN | |
| Primary | |
| Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title