Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OCEL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179396
Description
OCEL1 Polyclonal specifically detects OCEL1 in Human samples. It is validated for Western Blot.Specifications
OCEL1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FLJ22709, FWP009, occludin/ELL domain containing 1, occludin/ELL domain-containing protein 1, S863-9 | |
Rabbit | |
Affinity purified | |
RUO | |
79629 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_078854 | |
OCEL1 | |
Synthetic peptide directed towards the C terminal of human OCEL1The immunogen for this antibody is OCEL1. Peptide sequence VWREFEMKRMDPGFLDKQARCHYLKGKLRHLKTQIQKFDDQGDSEGSVYF. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Human: 100%; Pig: 92%; Guinea pig: 85%; Equine: 85%; Canine: 78%. | |
Human, Rat, Pig, Bovine, Canine, Equine, Guinea Pig | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction