Learn More
OCIAD1 Mouse anti-Human, Mouse, Rat, Clone: 1C10C3, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | OCIAD1 |
|---|---|
| Clone | 1C10C3 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7114
![]()
|
Proteintech Group Inc
666981IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody detects OCIAD1 with an apparent molecular weight of ∽35 kDa as has been demonstrated by several researches.
The Ras-related superfamily of guanine nucleotide binding proteins, which includes the R-Ras, Rap, Ral/Rec and Rho/Rab subfamilies, exhibit 30-60% homology with Ras p21. Accumulating data suggests an important role for Rab proteins, either in endocytosis or in biosynthetic protein transport. At least eight members of the Rab subfamily have been identified, each of which is found at a particular stage of a membrane transport pathway.Specifications
| OCIAD1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Asrij, FLJ20455, OCIA, OCIA domain containing 1, OCIAD1, TPA018 | |
| Ociad1 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 1C10C3 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Q5XIG4, Q9CRD0, Q9NX40 | |
| 289590, 54940, 68095 | |
| OCIAD1 Fusion Protein Ag9977 | |
| Primary | |
| -20°C | |
| OCIAD1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.