Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCIL/CLEC2d Antibody (166C1309) - Azide Free, Novus Biologicals™
SDP

Catalog No. NBP233226 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP233226 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP233226 Supplier Novus Biologicals Supplier No. NBP233226
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OCIL/CLEC2d Monoclonal specifically detects OCIL/CLEC2d in Mouse samples. It is validated for Western Blot.

Specifications

Antigen OCIL/CLEC2d
Applications Western Blot
Classification Monoclonal
Clone 166C1309
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml
Gene Alias CLAXLLT-1, C-type lectin domain family 2 member D, C-type lectin domain family 2, member D, C-type lectin related f, Lectin-like NK cell receptor, Lectin-like transcript 1, LLT1Osteoclast inhibitory lectin, OCILmember D
Gene Symbols CLEC2D
Host Species Mouse
Immunogen This antibody was developed by immunizing mice with a synthetic peptide containing amino acids 2-18 (CVTKASLPMLSPTGSPQ) of mouse OCIL, GenBank no. gb AAL37198.1.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 29121
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.