Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oct-3/4 Antibody (240408), PE, Novus Biologicals™
SDP

Catalog No. NB297008 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297008 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297008 Supplier Novus Biologicals Supplier No. FAB1759P

Rat Monoclonal Antibody

Oct-3/4 Monoclonal antibody specifically detects Oct-3/4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen Oct-3/4
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 240408
Conjugate PE
Dilution Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias OCT3,Oct-3,OCT4,Oct-4,OCT4,Oct3/4,OTF3,OTF-3,OTF4,POU domain protein,POU domain, class 5, transcription factor 1,POU5F1
Host Species Rat
Immunogen E. coli-derived recombinant human Oct-3/4, Met1-Asn265 (Met262Leu), Accession # Q01860
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5460
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.