Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oct-4A Antibody (03), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB383137 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383137 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383137 Supplier Novus Biologicals Supplier No. NBP312775UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Oct-4A Monoclonal antibody specifically detects Oct-4A in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Oct-4A
Applications Flow Cytometry
Classification Monoclonal
Clone 03
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias OCT3;Oct-3;OCT4;Oct-4;OCT4;Oct3/4;OTF3;OTF-3;OTF4;POU domain protein;POU domain, class 5, transcription factor 1;POU5F1
Host Species Mouse
Immunogen A synthetic peptide corresponding to the C-terminus of Human Oct-4A.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5460
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.