Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oct-4B Antibody (OTI7H1), FITC, Novus Biologicals™
SDP

Catalog No. NBP273141F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273141F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273141F Supplier Novus Biologicals Supplier No. NBP273141F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Oct-4B Monoclonal specifically detects Oct-4B in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Oct-4B
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI7H1
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Symbols POU5F1
Host Species Mouse
Immunogen Full length recombinant protein of human Oct-4B (NP_002692) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5460.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.