Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCT2 Antibody (OCT2/2137) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP275735 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275735 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275735 Supplier Novus Biologicals Supplier No. NBP275735
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

44471 Monoclonal specifically detects 44471 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.

Specifications

Antigen OCT2
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OCT2/2137
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with No Preservative
Gene Accession No. P09086
Gene Alias homeobox protein, Lymphoid-restricted immunoglobulin octamer-binding protein NF-A2, OCT2Oct-2, Octamer-binding protein 2, Octamer-binding transcription factor 2, OTF-2, OTF2POU domain class 2, transcription factor 2, POU class 2 homeobox 2, POU domain, class 2, transcription factor 2
Gene Symbols POU2F2
Host Species Mouse
Immunogen Recombinant fragment of human OCT2 protein (around aa 112-297) (exact sequence is proprietary) (Uniprot: P09086)
Molecular Weight of Antigen 60 kDa
Purification Method Protein A or G purified
Quantity 100 μg
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5452.0
Test Specificity OCT2 is a transcription factor of the POU homeo-domain family that binds to the Ig gene octamer sites, regulating B-cell-specific genes. OCT2 expression can be used as a marker of B-cell lineage and differentiation. Germinal center B-cells, mantle B-cells, monocytoid B-cells, and plasma cells show high level expression of OCT2. Additionally, mantle cell lymphoma, follicular lymphoma, marginal zone lymphoma, plasmacytoma, Burkitt lymphoma, diffuse large cell lymphoma, diffuse large B-cell lymphoma, Hodgkin lymphoma display increased expression of OCT2. Several studies of OCT2 expression have shown a low level expression in pre-B, T-cell, myelomonocytic, and epithelial cell lines, whereas all mature B-cell lines have high levels of expression. In spite of scanty evidence for OCT2 expression in T cells, it is believed that this factor participates in transcriptional regulation during T-cell activation.
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.