Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCT2 Rabbit anti-Human, Clone: OCT2/7073R, Novus Biologicals™
SDP

Catalog No. NB177443 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177443 20 μg
NB177442 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB177443 Supplier Novus Biologicals Supplier No. NBP31374320UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

OCT2 Monoclonal antibody specifically detects OCT2. in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen OCT2.
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OCT2/7073R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. P09086
Gene Alias homeobox protein, Lymphoid-restricted immunoglobulin octamer-binding protein NF-A2, OCT2Oct-2, Octamer-binding protein 2, Octamer-binding transcription factor 2, OTF-2, OTF2POU domain class 2, transcription factor 2, POU class 2 homeobox 2, POU domain, class 2, transcription factor 2
Host Species Rabbit
Immunogen Recombinant fragment of human OCT2 protein (around aa 112-297) (exact sequence is proprietary) (Uniprot: P09086)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5452
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.