Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octacosane, 99+%

Catalog No. AC129275000
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 630-02-4 | C28H58 | 394.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-02-4
Molecular Formula C28H58
Molecular Weight (g/mol) 394.77
MDL Number MFCD00009355
InChI Key ZYURHZPYMFLWSH-UHFFFAOYSA-N
Synonym n-octacosane, n-octcosane, unii-vff49836p8, ch3-ch2 26-ch3, octacosane, analytical standard, ccris 680, octacosane, n, octacosane, heptacosane, methyl, n-octacosane 10g
PubChem CID 12408
ChEBI CHEBI:32943
IUPAC Name octacosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 61.0°C to 64.0°C
Color White
Density 0.8067g/mL
Boiling Point 440.0°C
Flash Point 227°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)26CH3
Quantity 500 g
Beilstein 01,I,71
Specific Gravity 0.8067
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, chloroform
Formula Weight 394.76
Percent Purity 99+%
Physical Form Shiny Flakes or Powder
Chemical Name or Material Octacosane
Show More Show Less
EINECSNumber 211-125-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.