Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCTN1/SLC22A4 Antibody (892147), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB314932 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314932 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314932 Supplier Novus Biologicals Supplier No. FAB10985APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OCTN1/SLC22A4 Monoclonal specifically detects OCTN1/SLC22A4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen OCTN1/SLC22A4
Applications Flow Cytometry
Classification Monoclonal
Clone 892147
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias Ergothioneine transporter, ET transporter, ETT, MGC34546, MGC40524, OCTN1integral membrane transport protein, Organic cation/carnitine transporter 1, solute carrier family 22 (organic cation/ergothioneine transporter), member 4, solute carrier family 22 member 4, UT2H
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human SLC22A4, Accession # Q9H015
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6583
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.