Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCTN2/SLC22A5 Antibody (893753), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB297499 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297499 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297499 Supplier Novus Biologicals Supplier No. FAB11004A
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OCTN2/SLC22A5 Monoclonal specifically detects OCTN2/SLC22A5 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen OCTN2/SLC22A5
Applications Flow Cytometry
Classification Monoclonal
Clone 893753
Conjugate APC
Dilution Flow Cytometry
Formulation PBS
Gene Alias CDSP, FLJ46769, High-affinity sodium-dependent carnitine cotransporter, OCTN2high-affinity sodium dependent carnitine cotransporter, OCTN2VT, organic cation transporter 2, organic cation transporter 5, Organic cation/carnitine transporter 2, SCD, solute carrier family 22 (organic cation transporter), member 5, solute carrier family 22 (organic cation/carnitine transporter), member 5, solute carrier family 22 member 5
Host Species Mouse
Immunogen NS0 transfected with human SLC22A5, Accession # O76082
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6584
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.