Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ocular development associated gene Antibody (OTI1H1), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N273142A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273142A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273142A405 Supplier Novus Biologicals Supplier No. NBP273142AF405

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Ocular development associated gene Monoclonal specifically detects Ocular development associated gene in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Ocular development associated gene
Applications Western Blot
Classification Monoclonal
Clone OTI1H1
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ22489, GATA zinc finger domain containing 1, GATA zinc finger domain-containing protein 1, ocular development associated, Ocular development-associated gene protein, ODAGFLJ40695, RG083M05.2
Gene Symbols GATAD1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 68-269 of human Ocular development associated gene (NP_066990) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 57798.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.