Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ocular development associated gene Antibody (OTI1H1), PerCP, Novus Biologicals™
SDP

Catalog No. NB302175 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302175 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302175 Supplier Novus Biologicals Supplier No. NBP273142PCP

Mouse Monoclonal Antibody

Ocular development associated gene Monoclonal antibody specifically detects Ocular development associated gene in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Ocular development associated gene
Applications Western Blot
Classification Monoclonal
Clone OTI1H1
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias FLJ22489, GATA zinc finger domain containing 1, GATA zinc finger domain-containing protein 1, ocular development associated, Ocular development-associated gene protein, ODAGFLJ40695, RG083M05.2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 68-269 of human GATAD1 (NP_066990) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 57798
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.