Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ODF3L2 Antibody, Novus Biologicals™
SDP

Catalog No. NB436310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB436310 25 μL
NBP238113 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB436310 Supplier Novus Biologicals Supplier No. NBP23811325UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ODF3L2 Polyclonal specifically detects ODF3L2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ODF3L2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q3SX64
Gene Alias C19orf19, Chromosome 19 Open Reading Frame 19, Outer Dense Fiber Of Sperm Tails 3-Like 2, Outer Dense Fiber Protein 3-Like Protein 2
Gene Symbols ODF3L2
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: VASPAYSLVRRPSEAPPQDTSPGPIYFLDPKVTRFGRSCTPAYSMQGRAKSRGPEVTPGP
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284451
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.