Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ODR4/TTG1 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB051615 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB051615 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB051615 Supplier Novus Biologicals Supplier No. NBP298046G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ODR4/TTG1 Polyclonal specifically detects ODR4/TTG1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ODR4/TTG1
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 488
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias chromosome 1 open reading frame 27, FLJ20505, hODR-4, LAG1-interacting protein, odorant response abnormal 4, odr-4, ODR4, protein odr-4 homolog, Transactivated by transforming growth factor beta protein 1, TTG1, TTG1A
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ODR4/TTG1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54953
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.