Learn More
Sellstrom™ Odyssey II Cleanroom Goggles
Sterilize by autoclaving with steam or gamma radiation†
$53.40
Specifications
Lens Tint Clear
Frame Color Blue
Material Polycarbonate
View More Specs
Alert:
†Autoclaving is NOT recommended for goggles with anti-fog lenses (Cat. No. 19-822-925 and 19-823-051).
Products 2
Catalog Number Mfr. No. Ventilation Details Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Ventilation Details Lens Tint Price Quantity & Availability  
19-823-050
Encompass
View Documents
Sellstrom™
80231
Non-vent body Clear
Each for $53.40
Only null left
Edge
Add to Cart
19-822-925
View Documents
Sellstrom™
80230
Non-vent Clear N/A N/A N/A
Description
Description
  • For aseptic and cleanroom conditions; designed to be easily cleaned
  • Great panoramic view
  • High temperature, nonvented and top-vented body options available
  • UV-absorbing polycarbonate, durable antifog coating
  • Ultra comfortable, soft goggle body for a snug seal
  • Easily fits over prescription eyewear
  • Made of anallergic materials
  • Easy strap adjustment
Order Info
Replacement parts available (See Cat. No. 19-823-051).
Specifications
Specifications
Clear
Blue
Polycarbonate
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.