Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ODZ1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. p-7209396 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3551 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3551 Supplier Proteintech Group Inc Supplier No. 216961AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The teneurin transmembrane protein 1 (TENM1) is a member of a family of four neuronal cell surface proteins homologous to the Drosophila pair-rule gene Ten-m. TENM1 is expressed primarily in the developing central nervous system and may be proteolytically cleaved with the intracellular domain translocating to the nucleus. TENM1 is a direct target of the homeobox transcription factor EMX2, a transcription factor thought to be important for area specification in the developing cortex.

Specifications

Antigen ODZ1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TENM1
Gene Accession No. Q9UKZ4, Q9WTS4
Gene Alias ODZ1, ODZ3, Protein Odd Oz/ten m homolog 1, Ten 1, TEN M1, Tenascin M1, Teneurin 1, TNM, TNM1
Gene Symbols TENM1
Host Species Rabbit
Immunogen ODZ1 Fusion Protein Ag16289
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10178, 23963
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.