Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ OF 1
Selective BRPF1B and BRPF2 inhibitor
Specifications
Purity | 98% |
---|---|
Molecular Formula | 4-Bromo-N-(2,3-dihydro-6-methoxy-1,3-dimethyl-2-oxo-1 H-benzimidazol-5-yl)-2-methylbenzenesulfonamide |
CAS | 919973-83-4 |
Content And Storage | 4°C |
Product Type | OF 1 |
Description
Selective BRPF1B and BRPF2 bromodomain inhibitor (Kd values are 100 and 500 nM respectively). Exhibits 39-fold selectivity for BRPF1B and BRPF2 over BRD4. Accelerates FRAP recovery at 5 μM in a BRPF2 FRAP assay. Inhibits RANKL-induced differentiation of primary murine bone marrow cells and human primary monocytes into osteoclasts.Specifications
98% | |
919973-83-4 | |
OF 1 | |
OF 1 |
4-Bromo-N-(2,3-dihydro-6-methoxy-1,3-dimethyl-2-oxo-1 H-benzimidazol-5-yl)-2-methylbenzenesulfonamide | |
4°C | |
M.W. (Observed): 440.31 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title